N-Boc-4-carboxymethoxypiperidine ≥98%

Supplier: Ambeed
Warning

A257607-1G A257607-10G A257607-250MG A257607-5G
77601-072EA 52.28 USD
77601-072 77668-104 77668-106 77879-482
N-Boc-4-carboxymethoxypiperidine ≥98%
N-Boc-4-carboxymethoxypiperidine

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₅
MDL Number: MFCD02178987
CAS Number: 161948-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR