7-Cbz-2-oxo-7-aza-spiro[3.5]nonane ≥98%

Supplier: Ambeed
Warning

Synonyms: Benzyl 2-oxo-7-azaspiro[3.5]nonane-7-carboxylate

A257273-1G A257273-250MG A257273-5G
77612-464EA 84.46 USD
77612-464 77612-466 77612-468
7-Cbz-2-oxo-7-aza-spiro[3.5]nonane ≥98%
7-Cbz-2-oxo-7-aza-spiro[3.5]nonane

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₃
MW: 273.327 g/mol
MDL Number: MFCD10000572
CAS Number: 147610-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR