4-Julolidine boronic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-9-yl)boronic acid
A257214-100MG
A257214-250MG
77877-857EA
97
USD
77877-857
77877-858
4-Julolidine boronic acid ≥98%
4-Julolidine boronic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BNO₂ MW: 217.072 g/mol |
MDL Number:
MFCD03001757 CAS Number: 391248-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...