Myristoyl-Gly-OH ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-Tetradecanamidoacetic acid

A256865-1G A256865-250MG A256865-5G
77668-084EA 18 USD
77668-084 77668-086 77668-088
Myristoyl-Gly-OH ≥97%
Myristoyl-Gly-OH

Precaución: for Research Use Only

Formula: C₁₆H₃₁NO₃
MW: 285.428 g/mol
MDL Number: MFCD00155627
CAS Number: 14246-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR