2,5-Diiodopyridine ≥98%

Supplier: Ambeed
Warning

A256811-5G A256811-1G
77593-604EA 83.03 USD
77593-604 77672-290
2,5-Diiodopyridine ≥98%
2,5-Diiodopyridine

Precaución: for Research Use Only

Formula: C₅H₃I₂N
MW: 330.89
MDL Number: MFCD00234028
CAS Number: 116195-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR