2-Bromo-6-methoxypyrazine ≥98%

Supplier: Ambeed
Danger

A256548-250MG A256548-5G
77883-697EA 75 USD
77883-697 77883-698
2-Bromo-6-methoxypyrazine ≥98%
2-Bromo-6-methoxypyrazine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MDL Number: MFCD10688549
CAS Number: 91678-76-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR