4-(Pentafluorothio)aniline ≥97%

Supplier: Ambeed
Danger

Synonyms: 4-Aminophenylsulphur pentafluoride

A256402-1G A256402-250MG A256402-5G
77616-070EA 74.16 USD
77616-070 77748-476 77748-477
4-(Pentafluorothio)aniline ≥97%
4-(Pentafluorothio)aniline

Precaución: for Research Use Only

Formula: C₆H₆F₅NS
MW: 219.18 g/mol
MDL Number: MFCD01076203
CAS Number: 2993-24-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR