8-Methoxyquinoline-5-boronic acid ≥95%

Supplier: Ambeed
Warning

A255781-100MG A255781-1G A255781-250MG
77890-444EA 115 USD
77890-444 77890-445 77890-446
8-Methoxyquinoline-5-boronic acid ≥95%
8-Methoxyquinoline-5-boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₃
MDL Number: MFCD07357508
CAS Number: 1025735-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR