5-Bromo-2-(methylsulfonyl)benzoic acid ≥95%

Supplier: Ambeed
Warning

A255747-250MG A255747-1G
77612-454EA 230.72 USD
77612-454 77683-776
5-Bromo-2-(methylsulfonyl)benzoic acid ≥95%
5-Bromo-2-(methylsulfonyl)benzoic acid

Precaución: For research use only.

Formula: C₈H₇BrO₄S
MW: 279.10 g/mol
MDL Number: MFCD23701693
CAS Number: 146431-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR