Phthalocyanine ≥93%

Supplier: Ambeed
Warning

A255579-1G A255579-5G A255579-25G
77601-056EA 23.58 USD
77601-056 77601-058 77879-469
Phthalocyanine ≥93%
Phthalocyanine

Precaución: for Research Use Only

Formula: C₃₂H₁₈N₈
MDL Number: MFCD00005085
CAS Number: 574-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR