Methyl 1,3-benzoxazole-4-carboxylate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl benzo[d]oxazole-4-carboxylate
A255440-250MG
A255440-1G
77683-774EA
54
USD
77683-774
77683-773
Methyl 1,3-benzoxazole-4-carboxylate ≥98%
Methyl 1,3-benzoxazole-4-carboxylate
Precaución: For research use only.
Formula:
C₉H₇NO₃ MW: 177.157 g/mol |
MDL Number:
MFCD11110250 CAS Number: 128156-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...