4-Methyl-2,3-dihydro-1H-indole ≥95%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: 4-Methylindoline

A255416-1G A255416-250MG
77683-771EA 165 USD
77683-771 77683-772
4-Methyl-2,3-dihydro-1H-indole ≥95%, Ambeed.Inc
4-Methyl-2,3-dihydro-1H-indole

Precaución: For research use only.

Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD08437632
CAS Number: 62108-16-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR