Dicloxacillin sodium salt monohydrate ≥98%

Supplier: Ambeed
Danger

Synonyms: 3-(2,6-Dichlorophenyl)-5-methyl-4-isoxazolyl penicillin sodium salt monohydrate, Sodium (2S,5R,6R)-6-(3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate, Dicloxacillin Sodium hydrate

A255182-1MG A255182-50MG
77601-054EA 8.2 USD
77601-054 77626-340
Dicloxacillin sodium salt monohydrate ≥98%
Dicloxacillin sodium salt monohydrate

Precaución: for Research Use Only

Formula: C₁₉H₁₆Cl₂N₃NaO₅S·1H₂O
MW: 510.33 g/mol
MDL Number: MFCD00056865
CAS Number: 13412-64-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR