Bis-PEG7-acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 4,7,10,13,16,19,22-Heptaoxapentacosane-1,25-dioic acid, Bis-dPEG®7-acid

A254770-1G A254770-250MG
77616-064EA 181.28 USD
77616-064 77616-066
Bis-PEG7-acid ≥98%
Bis-PEG7-acid

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₁₁
MW: 426.46 g/mol
MDL Number: MFCD11041125
CAS Number: 94376-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR