4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)benzonitrile ≥95%

Supplier: Ambeed
Warning

A253616-100MG A253616-1G A253616-250MG A253616-5G
77892-471EA 70 USD
77892-471 77892-472 77892-473 77892-474
4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)benzonitrile ≥95%
4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₈BN₃O₂
MDL Number: MFCD21099763
CAS Number: 1402166-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR