(S)-N-(2-(2-Cyano-4,4-difluoropyrrolidin-1-yl)-2-oxoethyl)quinoline-4-carboxamide ≥97%

Supplier: Ambeed
Warning

Synonyms: SP-13786

A252113-1MG A252113-100MG A252113-10MG A252113-50MG A252113-5MG
77616-050EA 13.39 USD
77616-050 77746-401 77746-402 77746-403 77746-404
(S)-N-(2-(2-Cyano-4,4-difluoropyrrolidin-1-yl)-2-oxoethyl)quinoline-4-carboxamide ≥97%
(S)-N-(2-(2-Cyano-4,4-difluoropyrrolidin-1-yl)-2-oxoethyl)quinoline-4-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₁₄F₂N₄O₂
MW: 344.32 g/mol
MDL Number: MFCD29477499
CAS Number: 1448440-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR