5-(tert-Butyl)thiazol-2-amine ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A251696-250MG A251696-1G A251696-5G
77683-755EA 42 USD
77683-755 77890-422 77890-423
5-(tert-Butyl)thiazol-2-amine ≥98%, Ambeed.Inc
5-(tert-Butyl)thiazol-2-amine

Precaución: For research use only.

Formula: C₇H₁₂N₂S
MDL Number: MFCD02170355
CAS Number: 299417-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR