2-(Difluoromethyl)benzene boronic acid, pinacol ester ≥95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(2-(Difluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Difluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
A251633-250MG
A251633-100MG
A251633-1G
77683-754EA
160
USD
77683-754
77683-752
77683-753
2-(Difluoromethyl)benzene boronic acid, pinacol ester ≥95%
2-(Difluoromethyl)benzene boronic acid, pinacol ester
Precaución: For research use only.
Formula:
C₁₃H₁₇BF₂O₂ MW: 254.081 g/mol |
MDL Number:
MFCD16996279 CAS Number: 879275-72-6 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...