Pyridoxine ≥98%

Supplier: Ambeed
Warning

Synonyms: Vitamine B6 base, Pyridoxol, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol

A251506-10MG A251506-1MG A251506-50MG
77672-242EA 6 USD
77672-242 77672-244 77672-246
Pyridoxine ≥98%
Pyridoxine

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MW: 169.18 g/mol
MDL Number: MFCD00006335
CAS Number: 65-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR