[(4-Methyl-2-oxo-2H-chromen-7-yl)oxy]acetic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-((4-Methyl-2-oxo-2H-chromen-7-yl)oxy)acetic acid, 7-(Carboxymethoxy)-4-methylcoumarin

A251461-1G A251461-250MG
77626-322EA 160.68 USD
77626-322 77626-324
[(4-Methyl-2-oxo-2H-chromen-7-yl)oxy]acetic acid ≥95%
[(4-Methyl-2-oxo-2H-chromen-7-yl)oxy]acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₅
MW: 234.21 g/mol
MDL Number: MFCD00010854
CAS Number: 64700-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR