GSK-961081 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(3-((2-chloro-4-(((2-hydroxy-2-(8-hydroxy-2-oxo-1,2-dihydroquinolin-5-yl)ethyl)amino)methyl)-5-methoxyphenyl)amino)-3-oxopropyl)piperidin-4-yl [1,1'-biphenyl]-2-ylcarbamate, Batefenterol, TD-5959

A250343-1MG A250343-5MG
77748-470EA 11 USD
77748-470 77748-471
GSK-961081 98+%
GSK-961081

Precaución: for Research Use Only

Formula: C₄₀H₄₂ClN₅O₇
MDL Number: MFCD27987925
CAS Number: 743461-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR