(2-Morpholinopyridin-4-yl)boronic acid ≥96%

Supplier: Ambeed
Warning

A250050-100MG A250050-1G A250050-250MG
77881-662EA 69 USD
77881-662 77881-663 77881-664
(2-Morpholinopyridin-4-yl)boronic acid ≥96%
(2-Morpholinopyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BN₂O₃
MW: 208.02 g/mol
MDL Number: MFCD11054039
CAS Number: 952206-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR