Pyrazolo[1,5-a]pyrazine-3-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A249757-1G A249757-250MG A249757-100MG A249757-5G
77612-410EA 164.8 USD
77612-410 77685-705 77900-701 77901-633
Pyrazolo[1,5-a]pyrazine-3-carboxylic acid ≥95%
Pyrazolo[1,5-a]pyrazine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD11865260
CAS Number: 53902-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR