5-Chloromethyl-2-oxazolidinone ≥98%

Supplier: Ambeed
Danger

Synonyms: 5-(Chloromethyl)oxazolidin-2-one

A249350-1G A249350-5G
77593-572EA 18.45 USD
77593-572 77626-310
5-Chloromethyl-2-oxazolidinone ≥98%
5-Chloromethyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₄H₆ClNO₂
MW: 135.55 g/mol
MDL Number: MFCD00005269
CAS Number: 22625-57-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR