2-(tert-Butoxycarbonyl)-2-azaspiro[3.5]nonane-7-carboxylic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[(tert-butoxy)carbonyl]-2-azaspiro[3.5]nonane-7-carboxylic acid
A249021-250MG
A249021-100MG
A249021-1G
A249021-5G
77612-404EA
216.3
USD
77612-404
77685-702
77892-452
77892-453
2-(tert-Butoxycarbonyl)-2-azaspiro[3.5]nonane-7-carboxylic acid ≥98%
2-(tert-Butoxycarbonyl)-2-azaspiro[3.5]nonane-7-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₂₃NO₄ |
MDL Number:
MFCD22566156 CAS Number: 1363381-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...