[4-(1-Cyano-1-methylethyl)phenyl]boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Cyanoprop-2-yl)benzeneboronic acid, 2-(4-Boronophenyl)-2-methylpropanenitrile

A248969-100MG A248969-250MG A248969-5G
77872-240EA 35 USD
77872-240 77872-241 77872-242
[4-(1-Cyano-1-methylethyl)phenyl]boronic acid ≥98%
[4-(1-Cyano-1-methylethyl)phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BNO₂
MW: 189.02 g/mol
MDL Number: MFCD06659832
CAS Number: 850568-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR