(3R,4R)-tert-Butyl 4-amino-3-hydroxypiperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A248630-100MG A248630-250MG A248630-1G
77612-396EA 51.5 USD
77612-396 77612-398 77885-650
(3R,4R)-tert-Butyl 4-amino-3-hydroxypiperidine-1-carboxylate ≥98%
(3R,4R)-tert-Butyl 4-amino-3-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD18072074
CAS Number: 1007596-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR