4-(3-(Piperidin-1-yl)propoxy)phenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)propyl)piperidine

A248442-100MG A248442-1G A248442-250MG
77890-408EA 135 USD
77890-408 77890-409 77890-410
4-(3-(Piperidin-1-yl)propoxy)phenylboronic acid pinacol ester ≥98%
4-(3-(Piperidin-1-yl)propoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₃₂BNO₃
MDL Number: MFCD09475821
CAS Number: 401895-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR