2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

A248099-1G A248099-250MG A248099-5G
77672-218EA 49 USD
77672-218 77883-669 77883-670
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥98%
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BN₂O₃
MDL Number: MFCD08458483
CAS Number: 945863-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR