2-(Dimethylamino)-6-fluoronicotinaldehyde ≥97%

Supplier: Ambeed
Danger

A247679-1G A247679-250MG
77746-388EA 52 USD
77746-388 77746-389
2-(Dimethylamino)-6-fluoronicotinaldehyde ≥97%
2-(Dimethylamino)-6-fluoronicotinaldehyde

Precaución: for Research Use Only

Formula: C₈H₉FN₂O
MW: 168.17 g/mol
MDL Number: MFCD22577180
CAS Number: 193481-62-8
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR