(11aR)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-9-phenanthrenyl-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-oxide ≥98%
Supplier: Ambeed
|
Warning
|
A247428-100MG
A247428-1G
A247428-250MG
A247428-50MG
77746-384EA
207
USD
77746-384
77746-385
77746-386
77746-387
(11aR)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-9-phenanthrenyl-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-oxide ≥98%
(11aR)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-9-phenanthrenyl-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-oxide
Precaución: for Research Use Only
Formula:
C₄₅H₃₁O₄P MW: 666.7 g/mol |
MDL Number:
MFCD29905022 CAS Number: 1372719-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...