(1S,2R)-2-fluorocyclopropan-1-amine hydrochloride ≥98%

Supplier: Ambeed
Warning

A247306-250MG A247306-1G
77616-024EA 82.4 USD
77616-024 77748-447
(1S,2R)-2-fluorocyclopropan-1-amine hydrochloride ≥98%
(1S,2R)-2-fluorocyclopropan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇ClFN
MW: 111.55 g/mol
MDL Number: MFCD19207082
CAS Number: 141042-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR