5-(Bromomethyl)-benzofurazan ≥97%

Supplier: Ambeed
Danger

A247253-250MG A247253-1G
77877-827EA 47 USD
77877-827 77877-826
5-(Bromomethyl)-benzofurazan ≥97%
5-(Bromomethyl)-benzofurazan

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MDL Number: MFCD00174369
CAS Number: 32863-31-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR