(2S,3R,4R,5S,6R)-2-(3-((5-(4-Fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol hemihydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Canagliflozin hemihydrate

A247239-10MG A247239-5G A247239-10G A247239-250MG A247239-50MG
77685-695EA 5 USD
77685-695 77685-696 77901-308 77900-816 77901-941
(2S,3R,4R,5S,6R)-2-(3-((5-(4-Fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol hemihydrate ≥98%
(2S,3R,4R,5S,6R)-2-(3-((5-(4-Fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol hemihydrate

Precaución: for Research Use Only

Formula: C₄₈H₅₂F₂O₁₁S₂
MW: 907.05 g/mol
MDL Number: MFCD28975933
CAS Number: 928672-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR