Methyl-3,5-diaminobenzoate 98+%

Supplier: Ambeed
Warning

A247017-25G A247017-1G A247017-5G
77668-012EA 148.44 USD
77668-012 77879-449 77879-450
Methyl-3,5-diaminobenzoate 98+%
Methyl-3,5-diaminobenzoate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00041433
CAS Number: 1949-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR