9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-(methylamino)-1H-purin-6(9H)-one ≥95%

Supplier: Ambeed
Warning

Synonyms: N2-Methylguanosine

A246781-10MG A246781-1MG A246781-5MG
77600-986EA 252.15 USD
77600-986 77600-988 77600-990
9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-(methylamino)-1H-purin-6(9H)-one ≥95%
9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-(methylamino)-1H-purin-6(9H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₅O₅
MW: 297.26 g/mol
MDL Number: MFCD00057039
CAS Number: 2140-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR