MK-7655 ≥98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S,5R)-7-Oxo-2-(piperidin-4-ylcarbamoyl)-1,6-diazabicyclo[3.2.1]octan-6-yl hydrogen sulphate, [(2S,5R)-7-oxo-2-(piperidin-4-ylcarbamoyl)-1,6-diazabicyclo[3.2.1]octan-6-yl] hydrogen sulphate, Relebactam

A246160-100MG A246160-50MG
77685-690EA 443 USD
77685-690 77685-691
MK-7655 ≥98%
MK-7655

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₄O₆S
MDL Number: MFCD28502833
CAS Number: 1174018-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR