2,6-Difluoro-3-(3',5'-dimethoxybenzyloxy)phenylboronic acid ≥98%

Supplier: Ambeed
Warning

A246085-1G A246085-5G
77890-402EA 12 USD
77890-402 77890-403
2,6-Difluoro-3-(3',5'-dimethoxybenzyloxy)phenylboronic acid ≥98%
2,6-Difluoro-3-(3',5'-dimethoxybenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₅BF₂O₅
MDL Number: MFCD07369738
CAS Number: 849062-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR