cis-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A246077-1G A246077-100MG A246077-250MG
77616-020EA 249.26 USD
77616-020 77748-444 77748-445
cis-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid ≥97%
cis-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₈FNO₄
MW: 247.26 g/mol
MDL Number: MFCD28016232
CAS Number: 1628475-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR