2,8-Bis(9H-carbazol-9-yl)dibenzothiophene 95+%

Supplier: Ambeed
Warning

A245651-1G A245651-250MG
77900-566EA 60 USD
77900-566 77900-647
2,8-Bis(9H-carbazol-9-yl)dibenzothiophene 95+%
2,8-Bis(9H-carbazol-9-yl)dibenzothiophene

Precaución: for Research Use Only

Formula: C₃₆H₂₂N₂S
MDL Number: MFCD28291905
CAS Number: 913738-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR