[Co(Br2bpy)3](PF6)3 ≥95%

Supplier: Ambeed
Warning

A2454855-1G A2454855-250MG A2454855-5G
77598-358EA 261.38 USD
77598-358 77598-360 77877-821
[Co(Br2bpy)3](PF6)3 ≥95%
[Co(Br2bpy)3](PF6)3

Precaución: for Research Use Only

Formula: C₃₀H₁₈Br₆CoF₁₈N₆P₃
CAS Number: 2982842-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR