Ethyl 2-fluoro-6-hydroxybenzoate ≥95%

Supplier: Ambeed
Danger

A245105-1G A245105-250MG
77748-437EA 64 USD
77748-437 77748-438
Ethyl 2-fluoro-6-hydroxybenzoate ≥95%
Ethyl 2-fluoro-6-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MDL Number: MFCD00153207
CAS Number: 208708-03-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR