Di-mu-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II) ≥98%

Supplier: Ambeed
Warning

Synonyms: Chloro(2'-amino-1,1'-biphenyl-2-yl)palladium(II) dimer, Di-µ-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II)

A245006-1G A245006-250MG
77746-374EA 98 USD
77746-374 77746-375
Di-mu-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II) ≥98%
Di-mu-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II)

Precaución: for Research Use Only

Formula: C₂₄H₁₈Cl₂N₂Pd₂
MDL Number: MFCD22200542
CAS Number: 847616-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR