7-Chloroquinoline-5-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A244875-100MG A244875-1G A244875-250MG
77685-686EA 238 USD
77685-686 77901-903 77902-002
7-Chloroquinoline-5-carboxylic acid ≥98%
7-Chloroquinoline-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
MDL Number: MFCD28975165
CAS Number: 1936252-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR