(-)-Quinine hydrochloride dihydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate

A244479-1MG A244479-100G A244479-10G A244479-1G A244479-25G A244479-5G
77668-002EA 5 USD
77668-002 77879-432 77879-433 77879-434 77879-435 77879-436
(-)-Quinine hydrochloride dihydrate ≥98%
(-)-Quinine hydrochloride dihydrate

Precaución: for Research Use Only

Formula: C₂₀H₂₅ClN₂O₂·2H₂O
MDL Number: MFCD00151248
CAS Number: 6119-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR