3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)-3-pyrrolidinol hydrochloride, 3-(Trifluoromethyl)pyrrolidin-3-ol hydrochloride

A244464-10G A244464-5G
77892-439EA 690 USD
77892-439 77892-440
3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride ≥97%
3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClF₃NO
MDL Number: MFCD19982573
CAS Number: 1334147-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR