3-Amino-2-methoxybenzonitrile ≥95%

Supplier: Ambeed
Warning

A244261-100MG A244261-250MG
77900-889EA 48 USD
77900-889 77901-384
3-Amino-2-methoxybenzonitrile ≥95%
3-Amino-2-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD18389725
CAS Number: 725718-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR