Irinotecan hydrochloride trihydrate ≥98%

Supplier: Ambeed
Danger

A244220-1MG A244220-5G
77600-976EA 5.13 USD
77600-976 77881-647
Irinotecan hydrochloride trihydrate ≥98%
Irinotecan hydrochloride trihydrate

Precaución: for Research Use Only

Formula: C₃₃H₃₈N₄O₆·HCl·3H₂O
MDL Number: MFCD01765731
CAS Number: 136572-09-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR