trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: trans-4-[(Boc-amino)methyl]cyclohexanecarboxylic Acid, trans-4-(Boc-aminomethyl)cyclohexanecarboxylic Acid

A100805-5G A100805-10G A100805-1G A100805-25G
77570-074EA 25 USD
77570-074 77599-472 77876-780 77876-781
trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid ≥97%
trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
MDL Number: MFCD00673773
CAS Number: 27687-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR