Fmoc-N-amido-PEG24-acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-PEG24-acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28,31,34,37,40,43,46,49,52,55,58,61,64,67,70,73,76-pentacosaoxa-4-azanonaheptacontan-79-oic acid

A1006777-100MG A1006777-250MG A1006777-1G
77619-648EA 112.27 USD
77619-648 77619-650 77782-824
Fmoc-N-amido-PEG24-acid ≥97%
Fmoc-N-amido-PEG24-acid

Precaución: for Research Use Only

Formula: C₆₆H₁₁₃NO₂₈
MDL Number: MFCD11041159
CAS Number: 2170484-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR